Anti-CHTF18 Rabbit Polyclonal Antibody

Supplier: Proteintech
13030-1-AP-150UL
10084-778EA 462.47 USD
10084-778
Anti-CHTF18 Rabbit Polyclonal Antibody
Antibodies
Western Blot:Y79 Cells, 1:200-1:2000; IHC: Human medulloblastoma, 1:20-1:200

Type: Primary
Antigen: CHTF18
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR