Anti-CHRNB1 Rabbit Polyclonal Antibody

Supplier: Proteintech
11553-1-AP-150UL
10084-748EA 462.47 USD
10084-748
Anti-CHRNB1 Rabbit Polyclonal Antibody
Antibodies
Acetylcholine receptors (AChRs) are integral membrane proteins that respond to the binding of acetylcholine (ACh), a neurotransmitter synthesized, stored and finally released by cholinergic neurons. After binding acetylcholine, the AChR responds by an extensive change in conformation that affects all subunits and leads to opening of an ion-conducting channel across the plasma membrane. The muscle acetylcholine receptor is composed of five subunits: two alpha subunits and one each of the beta, delta, and gamma (in immature muscle) or epsilon (in mature muscle) subunits. CHRNB1 gene encodes acetylcholine receptor subunit beta (ACHRB). Mutations in this gene are associated with slow-channel congenital myasthenic syndrome.

Western Blot: Human Liver Tissue, 1:200-1:2000; IHC: Human Colon cancer Tissue, 1:20-1:200

Type: Primary
Antigen: CHRNB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR