Anti-CHRNA5 Rabbit Polyclonal Antibody

Supplier: Proteintech
13516-1-AP-150UL
10084-742EA 462.47 USD
10084-742
Anti-CHRNA5 Rabbit Polyclonal Antibody
Antibodies
Western Blot:Jurkat Cells, 1:200-1:2000

Type: Primary
Antigen: CHRNA5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR