Anti-CHRNA10 Rabbit Polyclonal Antibody

Supplier: Proteintech
55291-1-AP-150UL
10084-738EA 462.47 USD
10084-738
Anti-CHRNA10 Rabbit Polyclonal Antibody
Antibodies
CHRNA10, also named as NACHRA10, belongs to the ligand-gated ion channel (TC 1.A.9) family, Acetylcholine receptor (TC 1.A.9.1) subfamily and Alpha-10/CHRNA10 sub-subfamily. It is an ionotropic receptor with a probable role in the modulation of auditory stimuli. In the ear, CHRNA10 may lead to a reduction in basilar membrane motion, altering the activity of auditory nerve fibers and reducing the range of dynamic hearing. It may protect against acoustic trauma. This antibody is specific to CHRNA10.

Western Blot: Raji Cells, 1:200-1:1000

Type: Primary
Antigen: CHRNA10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR