Anti-CHRAC1 Rabbit Polyclonal Antibody

Supplier: Proteintech
12126-2-AP-150UL
10084-734EA 462.47 USD
10084-734
Anti-CHRAC1 Rabbit Polyclonal Antibody
Antibodies
Western Blot:K-562 Cells, 1:200-1:1000; IHC: Human pancreas cancer, 1:20-1:200

Type: Primary
Antigen: CHRAC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR