Anti-CHMP4B Rabbit Polyclonal Antibody

Supplier: Proteintech
13683-1-AP-150UL
10084-720EA 462.47 USD
10084-720
Anti-CHMP4B Rabbit Polyclonal Antibody
Antibodies
CHMP4B (charged multivesicular body protein 4b) is a human homologue of yeast Snf7p (also named Vps32p) and a member of CHMP family which is implicated in multivesicular body (MVB) sorting. The CHMP proteins are components of ESCRT-III (endosomal sorting complex required for transport III), a complex involved in degradation of surface receptor proteins and formation of endocytic multivesicular bodies. Among seven CHMP family members, only CHMP4 has been found to interact with Alix, a multifunctional adaptor protein in various cellular functions. This antibody is highly specific to the CHMP4B and it doesn’t cross-react with other CHMPs as well as other CHMP4s (CHMP4A and CHMP4C).

Western Blot: Mouse Heart Tissue, 1:200-1:1000; IHC: Human oesophagus cancer Tissue, 1:20-1:200

Type: Primary
Antigen: CHMP4B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR