Anti-CHMP1A Rabbit Polyclonal Antibody
Supplier: Proteintech
15761-1-AP-150UL
10084-712EA 462.47 USD
10084-712
Anti-CHMP1A Rabbit Polyclonal Antibody
Antibodies
Charged multivesicular body protein 1A (CHMP1A), also known as chromatin modifying protein 1A, is a member of the ESCRT-III (endosomal sorting complex required for transport III) complex which is involved in degradation of internalized transmembrane receptor proteins and formation of multivesicular bodies (MVBs) . CHMP1A has both nuclear and cytoplasmic distributions . Subcellular localization of CHMP1A seems to vary depending on the cell type . CHMP1A is required for MVBs formation and has a role in stable gene silencing within the nucleus. Besides, the tumor suppressor role of CHMP1A hasMore Product Information