Anti-CHFR Rabbit Polyclonal Antibody

Supplier: Proteintech
12169-1-AP-150UL
10084-700EA 462.47 USD
10084-700
Anti-CHFR Rabbit Polyclonal Antibody
Antibodies
The Checkpoint with forkhead and RING finger domains protein(CHFR) is a checkpoint protein that reportedly initiates a cell cycle delay in response to microtubule stress during prophase in mitosis and it has been hypothesized to be a tumor suppressor with a potential role as a biomarker for chemotherapeutic response to Taxol. It belongs to the CHFR family. Endogenous Chfr is ubiquitinated and phosphorylated in an MPF-dependent manner in mitotic Xenopus egg extracts. It has 5 isoforms produced by alternative splicing.

Western Blot: HepG2 Cells, 1:500-1:5000; IHC: Human endometrial cancer Tissue, 1:20-1:200

Type: Primary
Antigen: CHFR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR