Anti-CFB Mouse Monoclonal Antibody [clone: 8D2B6]

Supplier: Proteintech
66154-1-IG-150UL
10084-684EA 462.47 USD
10084-684
Anti-CFB Mouse Monoclonal Antibody [clone: 8D2B6]
Antibodies
Complement factor B (CFB) is a component of the alternative pathway of complement activation. Complement factor B is cleaved into Ba and Bb by factor D in the presence of C3b. The active subunit Bb is a serine protease which associates with C3b to form the alternative pathway C3 convertase. Bb is involved in the proliferation of preactivated B lymphocytes, while Ba inhibits their proliferation.

Western Blot: Human Blood Tissue, 1:500-1:5000; IHC: Human Liver Tissue, 1:20-1:200

Type: Primary
Antigen: CFB
Clonality: Monoclonal
Clone: 8D2B6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR