Anti-CES3 Rabbit Polyclonal Antibody

Supplier: Proteintech
14587-1-AP-150UL
10084-678EA 462.47 USD
10084-678
Anti-CES3 Rabbit Polyclonal Antibody
Antibodies
CES3 is involved in the detoxification of xenobiotics and in the activation of ester and amide prodrugs. It is expressed in liver, colon and small intestine. This antibody recognizes both isoforms with MW 60-62 kDa and 24 kDa.

Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human Colon cancer Tissue, 1:20-1:200

Type: Primary
Antigen: CES3
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR