Anti-DEDD Rabbit Polyclonal Antibody

Supplier: Proteintech
10816-1-AP-150UL
10084-632EA 462.47 USD
10084-632
Anti-DEDD Rabbit Polyclonal Antibody
Antibodies
IHC: Human prostate cancer, 1:20-1:200

Type: Primary
Antigen: DEDD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR