Anti-BPIL1 Rabbit Polyclonal Antibody

Supplier: Proteintech
13461-2-AP-150UL
10083-658EA 462.47 USD
10083-658
Anti-BPIL1 Rabbit Polyclonal Antibody
Antibodies
Bactericidal/permeability-increasing protein-like 1 (BPIL1, also known as LPLUNC2 or BPIFB2) is a member of the lipid transfer (LT)/lipopolysaccharide binding protein (LBP) family. LT/LBP proteins are structurally related proteins capable of binding phospholipids and lipopolysaccharides . The gene of BPIL1 maps to Chromosome 20q11, and encodes a 458-amino acid protein with a calculated molecular weight of 49 kDa. It is highly expressed in hypertrophic tonsils. BPIL1 has been identified as a nasal mucous protein which may be involved in immune response in the nose against microbial infections . Reduced expression of antimicrobial PLUNC proteins (LPLUNC1 and LPLUNC2) has been reported in nasal polyp tissues of patients with chronic rhinosinusitis .

Western Blot:A549 Cells, 1:500-1:5000; IHC: Human lung cancer, 1:20-1:200

Type: Primary
Antigen: BPIL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR