Anti-BMPR2 Rabbit Polyclonal Antibody

Supplier: Proteintech
14376-1-AP-150UL
10083-638EA 462.47 USD
10083-638
Anti-BMPR2 Rabbit Polyclonal Antibody
Antibodies
BMPs (bone morphogenetic protein) are involved in endochondral bone formation and embryogenesis. BMPR2 encodes a member of the (BMP) receptor family of transmembrane serine/threonine kinases. It is a widely expressed receptor, with high mRNA expression during development in many tissues. Dimer of BMPR2 receptors (70-80 kD) forms a complex with a dimer of type I BMPR1(50-55 kD) in signal transduction. This antibody recognizes endogenous levels of total BMPR2 protein, including 115 kD precursor, 75 kD mature form and 150 kD dimer.

Western Blot: Mouse Brain Tissue, 1:500-1:5000; IP: Mouse Brain Tissue, 1:200-1:2000

Type: Primary
Antigen: BMPR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR