Anti-cIAP2 Rabbit Polyclonal Antibody

Supplier: Proteintech
24304-1-AP-150UL
10083-612EA 462.47 USD
10083-612
Anti-cIAP2 Rabbit Polyclonal Antibody
Antibodies
Western Blot:U-937 Cells, 1:500-1:5000; IHC: Human tonsillitis Tissue, 1:20-1:200

Type: Primary
Antigen: cIAP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR