Anti-BCL2 Mouse Monoclonal Antibody [clone: 4H8C6]

Supplier: Proteintech
60178-1-IG-150UL
10083-550EA 462.47 USD
10083-550
Anti-BCL2 Mouse Monoclonal Antibody [clone: 4H8C6]
Antibodies
BCL2 belongs to the Bcl-2 family. It is a suppresses apoptosis in a variety of cell systems including factor-dependent lymphohematopoietic and neural cells. BCL2 regulates cell death by controlling the mitochondrial membrane permeability. It appears to function in a feedback loop system with caspases. BCL2 inhibits caspase activity either by preventing the release of cytochrome c from the mitochondria and/or by binding to the apoptosis-activating factor (APAF-1).

Western Blot:Jurkat Cells, 1:500-1:5000; FC:Jurkat Cells, N/A

Type: Primary
Antigen: BCL2
Clonality: Monoclonal
Clone: 4H8C6
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR