Anti-BAP31 Rabbit Polyclonal Antibody

Supplier: Proteintech
11200-1-AP-150UL
10083-504EA 462.47 USD
10083-504
Anti-BAP31 Rabbit Polyclonal Antibody
Antibodies
Western Blot: HEK-293 Cells, 1:1000-1:10000; IHC: Human Colon cancer Tissue, 1:20-1:200; IF: Hela Cells, 1:10-1:100; IP:A431 Cells, 1:500-1:5000; FC: HeLa Cells, N/A

Type: Primary
Antigen: BAP31
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR