Anti-C18orf55 Rabbit Polyclonal Antibody

Supplier: Proteintech
16641-1-AP-150UL
10083-304EA 462.47 USD
10083-304
Anti-C18orf55 Rabbit Polyclonal Antibody
Antibodies
IHC: Human Liver, 1:20-1:200

Type: Primary
Antigen: C18orf55
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR