Anti-ARHGAP1 Rabbit Polyclonal Antibody

Supplier: Proteintech
11169-1-AP-150UL
10083-066EA 462.47 USD
10083-066
Anti-ARHGAP1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human pancreas Tissue, 1:200-1:1000; IHC: Human pancreas cancer Tissue, 1:20-1:200

Type: Primary
Antigen: ARHGAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR