Anti-ARFGAP3 Rabbit Polyclonal Antibody

Supplier: Proteintech
15293-1-AP-150UL
10083-052EA 462.47 USD
10083-052
Anti-ARFGAP3 Rabbit Polyclonal Antibody
Antibodies
Western Blot: HepG2 Cells, 1:1000-1:10000; IF:A431 Cells, 1:10-1:100; IHC: Human breast cancer Tissue, 1:20-1:200

Type: Primary
Antigen: ARFGAP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR