Anti-ANO1 Rabbit Polyclonal Antibody

Supplier: Proteintech
12652-1-AP-150UL
10082-938EA 462.47 USD
10082-938
Anti-ANO1 Rabbit Polyclonal Antibody
Antibodies
ANO1 also named as DOG1, ORAOV2, TAOS2 and TMEM16A, acts as a calcium-activated chloride channel. It is required for normal tracheal development. ANO1 (or DOG1) is used as an aid in the identification and diagnosis of gastrointestinal stromal tumors (GIST) within the context of an antibody panel, the patient’s clinical history, and other diagnostic tests evaluated by a qualified pathologist. The immunogen is the C-terminal of ANO1. This antibody can recognize isoform1 and isoform2 of human ANO1.

IHC: Human Colon cancer Tissue, 1:20-1:200

Type: Primary
Antigen: ANO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR