Anti-ANXA5 Rabbit Polyclonal Antibody

Supplier: Proteintech
11060-1-AP-150UL
10082-930EA 462.47 USD
10082-930
Anti-ANXA5 Rabbit Polyclonal Antibody
Antibodies
Western Blot: HeLa Cells, 1:200-1:2000; IHC: Human spleen Tissue, 1:20-1:200; IP: HeLa Cells, 1:200-1:2000; FC: HeLa Cells, N/A; IF: HeLa Cells, 1:10-1:100

Type: Primary
Antigen: ANXA5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR