Anti-ANKS1B Rabbit Polyclonal Antibody

Supplier: Proteintech
24783-1-AP-150UL
10082-914EA 462.47 USD
10082-914
Anti-ANKS1B Rabbit Polyclonal Antibody
Antibodies
ANKS1B also named as AIDA or cajalin 2 is a amino acid protein, which contains 7 ANK repeats, 1 PID domain and 2 SAM domians. ANKS1B localizes in cytoplasm and is highly expressed in brain and testis. ANKS1B exists as many alternatively spliced isoforms. It interacts with AbPP to play a role in brain development. AIDA-1 also interacts with coilin in Cajal bodies to regulate pre-mRNA splicing.

Western Blot:A549 Cells, 1:500-1:5000; IF: HepG2 Cells, 1:20-1:200; IHC: Human Testis Tissue, 1:20-1:200

Type: Primary
Antigen: ANKS1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR