Anti-ANKRD55 Rabbit Polyclonal Antibody

Supplier: Proteintech
24203-1-AP-150UL
10082-910EA 462.47 USD
10082-910
Anti-ANKRD55 Rabbit Polyclonal Antibody
Antibodies
ANKRD55 also named as Ankyrin repeat domain containing protein 55 is a 614 amino acid protein, which contains 9 ANK repeat. ANKRD55 may exist three alternatively spliced isoforms and belongs to ankyrin family. The funciton of ANKRD55 is still non-known and need to be further studied.

Western Blot:L02 Cells, 1:500-1:5000; IHC: Human cervical cancer Tissue, 1:20-1:200

Type: Primary
Antigen: ANKRD55
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR