Anti-ANGPTL2 Rabbit Polyclonal Antibody

Supplier: Proteintech
12316-1-AP-150UL
10082-884EA 462.47 USD
10082-884
Anti-ANGPTL2 Rabbit Polyclonal Antibody
Antibodies
ANGPTL2 (angiopoietin-like 2), a member of the Angptl protein family, is predominantly secreted from adipose tissue and the heart. ANGPTL2 consists of an N-terminus with a conserved coiled-coil domain, two glycosylation sites and a C-terminus with a conserved fibrinogen-like domain. It is a key adipocyte-derived inflammatory mediator that links obesity to systemic insulin resistance. ANGPTL2 may play an important role in the pathogenesis of diabetic glomerulopathy.

Western Blot: Mouse small intestine Tissue, 1:500-1:5000; IHC: Human Heart Tissue, 1:20-1:200

Type: Primary
Antigen: ANGPTL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR