Anti-AMY2A Rabbit Polyclonal Antibody

Supplier: Proteintech
15845-1-AP-150UL
10082-868EA 462.47 USD
10082-868
Anti-AMY2A Rabbit Polyclonal Antibody
Antibodies
The complete human amylase gene contains five intact genes: two pancreatic genes (AMY2A and AMY2B) and three salivary genes (AMYIA, AMYIB, and AMYIC).AMY2A(Pancreatic alpha-amylase)is also named as PA and is a 56 kDa protein consisting of 496 amino acids in a single polypeptide chain, folded into three domains.AMY2A and AMY2B transcripts are present in the human pancreas but not in the parotid salivary gland, while AMY1 transcripts are present in the parotid gland but not in the pancreas.This antibody is specific to AMY2A.

Western Blot: Human Liver Tissue, 1:500-1:5000; IHC: Human pancreas, 1:20-1:200

Type: Primary
Antigen: AMY2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR