Anti-AMPD1 Rabbit Polyclonal Antibody

Supplier: Proteintech
19780-1-AP-150UL
10082-850EA 462.47 USD
10082-850
Anti-AMPD1 Rabbit Polyclonal Antibody
Antibodies
AMPD1, also named as MAD and MADA, belongs to the adenosine and AMP deaminases family. AMP deaminase plays a critical role in energy metabolism. It catalyzes the reaction: AMP + H2O = IMP + NH3. Defects in AMPD1 are the cause of adenosine monophosphate deaminase deficiency muscle type (AMPDDM) which is a metabolic disorder resulting in exercise-related myopathy. The antibody is specific to AMPD1.

Western Blot: Mouse Skeletal Muscle Tissue, 1:500-1:5000

Type: Primary
Antigen: AMPD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR