Anti-AMMECR1 Rabbit Polyclonal Antibody

Supplier: Proteintech
24687-1-AP-150UL
10082-840EA 462.47 USD
10082-840
Anti-AMMECR1 Rabbit Polyclonal Antibody
Antibodies
AMMECR1 also named as AMME syndrome candidate gene 1 protein is a 333 amino acid protein, which contains 1 AMMECER1 domain. Containing a glycine-rich N terminus, the AMMECR1 protein exhibits putative nuclear localization and a substantial level of instability, suggesting it plays a role in regulation. The deletion of AMMECR1 gene may be involved in glomerulonephritis, sensorineural hearing loss, mental retardation, midface hypoplasia and elliptocytosis.

Western Blot:Jurkat Cells, 1:200-1:2000; IHC: Human cerebellum Tissue, 1:20-1:200

Type: Primary
Antigen: AMMECR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR