Anti-ALX3 Rabbit Polyclonal Antibody

Supplier: Proteintech
22358-1-AP-150UL
10082-824EA 462.47 USD
10082-824
Anti-ALX3 Rabbit Polyclonal Antibody
Antibodies
ALX3 is a transcriptional regulator with a possible role in patterning of mesoderm during development. It is a paired-class aristaless-like homeoprotein expressed in different tissues during embryonic development. We always got 70 kDa dimer form in our detection.

Western Blot: Mouse Testis Tissue, 1:500-1:5000

Type: Primary
Antigen: ALX3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR