Anti-ALG6 Rabbit Polyclonal Antibody

Supplier: Proteintech
20753-1-AP-150UL
10082-790EA 462.47 USD
10082-790
Anti-ALG6 Rabbit Polyclonal Antibody
Antibodies
Western Blot: HeLa Cells, 1:200-1:2000

Type: Primary
Antigen: ALG6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR