Anti-ALDH1A2 Rabbit Polyclonal Antibody

Supplier: Proteintech
13951-1-AP-150UL
10082-754EA 462.47 USD
10082-754
Anti-ALDH1A2 Rabbit Polyclonal Antibody
Antibodies
ALDH1A2(aldehyde dehydrogenase family 1 member A2) is also named as RALDH2(retinal dehydrogenase 2) and belongs to the aldehyde dehydrogenase family. It is a cytosolic homotetramer(56.7 kDa subunits) exhibiting complex expression patterns throughout embryonic development. ALDH1A2 plays a role in retinoid metabolism during embryonic development where it is considered to be the major retinoic acid-synthesizing enzyme during early embryogenesis. Its expression is reduced in primary prostate tumors compared with normal prostate tissue suggested that it is normally expressed in the epithelial compartment of prostate tissue.It has 2 isoforms produced by alternative splicing.

Western Blot: Mouse Testis Tissue, 1:500-1:5000

Type: Primary
Antigen: ALDH1A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR