Anti-IGFBP2 Rabbit Polyclonal Antibody

Supplier: Proteintech
11065-3-AP-150UL
10082-156EA 462.47 USD
10082-156
Anti-IGFBP2 Rabbit Polyclonal Antibody
Antibodies
Insulin-like growth factors (IGFs) and their binding proteins (IGFBPs) have important metabolic roles in the human body. IGFBP-2 is a 34 kD IGFBP protein with degraded fragments of 24 kD, 19kd and 15kd. The levels of IGFBP2 are elevated during progression of many human cancers.

Western Blot: MCF7 Cells, 1:200-1:2000; IHC: Human prostate cancer, 1:20-1:200

Type: Primary
Antigen: IGFBP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR