Anti-ARF1 Rabbit Polyclonal Antibody

Supplier: Proteintech
10790-1-AP-150UL
10082-086EA 462.47 USD
10082-086
Anti-ARF1 Rabbit Polyclonal Antibody
Antibodies
ADP-ribosylation factors (ARFs) are members of the ARF family of GTP-binding proteins of the Ras superfamily, with 20kda protein size. ARFs bind and regulate GTP/GDP cycle by alternating between the active GTP-bound and inactive GDP-bound conformations. ARF family proteins are essential and ubiquitous in eukaryotes. Six highly conserved members of the family have been identified in mammalian cells. They function in vesicular traffic and actin remodelling and other bioprocesses in cells. The ARF1 protein is localized to the Golgi apparatus and has a central role in intra-Golgi transport. It mainly functions in coat recruitment. .This antibody can bind ARFs for the close sequences.

Western Blot: HeLa Cells, 1:200-1:2000; IHC: Human small intestine Tissue, 1:20-1:200

Type: Primary
Antigen: ARF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR