Anti-CaMKII beta Rabbit Polyclonal Antibody

Supplier: Proteintech
11533-1-AP-150UL
10081-986EA 462.47 USD
10081-986
Anti-CaMKII beta Rabbit Polyclonal Antibody
Antibodies
CAMK2B(Calcium/calmodulin-dependent protein kinase type II subunit beta ), also named as CAM2, CAMK2 and CAMKB, belongs to the protein kinase superfamily, CAMK Ser/Thr protein kinase family and CaMK subfamily. It is a prominent kinase in the central nervous system that may function in long-term potentiation and neurotransmitter release. CAMK2B is a member of the NMDAR signaling complex in excitatory synapses, it may regulate NMDAR-dependent potentiation of the AMPAR and synaptic plasticity. It plays a distinct role in the induction of anergy in T lymphocytes, by differential regulation of IL10 and IL2 gene transcription suggesting MEF2A as a molecular target which can integrate different calcium signals. This protein has 8 isoforms produced by alternative splicing with the molecular weight between 50 kDa and 73 kDa.

Western Blot: Mouse Brain Tissue, 1:500-1:5000; IHC: Human gliomas Tissue, 1:20-1:200

Type: Primary
Antigen: CaMKII beta
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR