Anti-BCL10 Rabbit Polyclonal Antibody

Supplier: Proteintech
50180-1-AP-150UL
10081-972EA 462.47 USD
10081-972
Anti-BCL10 Rabbit Polyclonal Antibody
Antibodies
The gene encoding BCL10 was identified by its recurrent translocation t(1;14)(p22;q32) in a case of mucosa-associated lymphoid tissue (MALT) lymphoma. BCL10 contains a caspase recruitment domain (CARD), and has been shown to induce apoptosis and to activa

  • Western Blot: Mouse ovary Tissue, 1:200-1:2000; IHC: Human lung cancer, 1:20-1:200

Type: Primary
Antigen: BCL10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR