Anti-ANXA2 Mouse Monoclonal Antibody [clone: 3H1B11]

Supplier: Proteintech
60051-1-IG-150UL
10081-934EA 462.47 USD
10081-934
Anti-ANXA2 Mouse Monoclonal Antibody [clone: 3H1B11]
Antibodies
Annexin A2 (ANXA2), also named Annexin-2 or Lipocortin II, is a Ca2+ binding protein that is up-regulated in virally transformed cell lines and human tumors. This calcium-regulated membrane-binding protein, whose affinity for calcium is greatly enhanced by anionic phospholipids, may cross-link plasma membrane phospholipids with actin and the cytoskeleton and be involved with exocytosis. Heat-stress response may also involve Annexin-2.

Western Blot:K-562 Cells, 1:500-1:5000; IHC: Human breast cancer Tissue, 1:20-1:200; IF: Hela Cells, 1:10-1:100; IP: HeLa Cells, 1:500-1:5000; FC: HeLa Cells, N/A

Type: Primary
Antigen: ANXA2
Clonality: Monoclonal
Clone: 3H1B11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR