Anti-ALDH1A1 Rabbit Polyclonal Antibody
Supplier: Proteintech
15910-1-AP-150UL
10081-918EA 462.47 USD
10081-918
Anti-ALDH1A1 Rabbit Polyclonal Antibody
Antibodies
ALDH1A1(Aldehyde dehydrogenase family 1 member A1 ), also named as ALDC, ALDH1 and PUMB1, belongs to the aldehyde dehydrogenase family. The ALDH1A1 gene encodes a liver cytosolic isoform of acetaldehyde dehydrogenase, an enzyme involved in the major pathway of alcohol metabolism after alcohol dehydrogenase. ALDH1A1 plays a critical role in protection against oxidative stress-induced cytotoxicity in lens epithelial cells. And it is important for multiple biological activities including drug resistance, cell differentiation, and oxidative stress response. As a novel cancer stem cell marker,More Product Information