Anti-HSF1 Rabbit Polyclonal Antibody

Supplier: Proteintech
16107-1-AP-150UL
10081-862EA 462.47 USD
10081-862
Anti-HSF1 Rabbit Polyclonal Antibody
Antibodies
HSF1 belongs to heat-shock transcription factor that activate heat-shock response genes under conditions of heat or other stresses. Also, HSF1 has linked with oogenesis, spermatogenesis, and placental development. It can activate AKT and inactivate JNK and CASP3 to protect cardiomyocytes from death. And it has a role in the regulation of life span and establishs a role for SIRT1 in protein homeostasis and heat-shock response.

Western Blot:K-562 Cells, 1:200-1:2000; IHC: Human lung cancer Tissue, 1:20-1:200

Type: Primary
Antigen: HSF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR