Anti-HNRNPU Rabbit Polyclonal Antibody

Supplier: Proteintech
16365-1-AP-150UL
10081-844EA 462.47 USD
10081-844
Anti-HNRNPU Rabbit Polyclonal Antibody
Antibodies
Heterogeneous nuclear ribonucleoproteins (hnRNPs), a major group of pre-mRNA-binding proteins, associate with nascent RNA polymerase II transcripts to form hnRNP complexes that participate in various steps of mRNA processing as well as the export, translation and stability of mRNAs. Protein hnRNPU, another hnRNPs family member, is able to stabilize specific mRNAs by binding to their 3′-untranslated regions (UTRs). Moreover, hnRNPU has a regulatory function during the initial phases of transcription activation by affecting the activity of RNA Pol II .

Western Blot: HEK-293 Cells, 1:1000-1:10000; IP: HeLa Cells, 1:200-1:2000; IHC: Human Liver Tissue, 1:20-1:200

Type: Primary
Antigen: HNRNPU
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR