Anti-HLA-F Rabbit Polyclonal Antibody

Supplier: Proteintech
14670-1-AP-150UL
10081-834EA 462.47 USD
10081-834
Anti-HLA-F Rabbit Polyclonal Antibody
Antibodies
Human major histocompatibility complex (MHC) antigens, also referred to as human leukocyte antigens (HLA), are encoded by genes located on the short arm of chromosome 6 (6p21.3). There are two classes of HLA antigens: class I and class II. This class I molecules are membrane glycoproteins composed of a heavy (alpha) chain which is encoded by a HLA class I gene, and β2-microglobulin light (beta) chain. The most extensively characterized members of the HLA class I gene family are the genes encoding the major transplantation antigenes, HLA-A, B and C. HLA-F is a non-classical MHC class I molecule.

Western Blot: Human Placenta Tissue, 1:200-1:2000; IHC: Human skin cancer Tissue, 1:20-1:200

Type: Primary
Antigen: HLA-F
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR