Anti-EVI2A Rabbit Polyclonal Antibody

Supplier: Proteintech
51014-2-AP-150UL
10081-764EA 462.47 USD
10081-764
Anti-EVI2A Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse Liver Tissue, 1:500-1:5000

Type: Primary
Antigen: EVI2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR