Anti-EEF1D Mouse Monoclonal Antibody [clone: 3G4C4]

Supplier: Proteintech
60085-2-IG-150UL
10081-744EA 462.47 USD
10081-744
Anti-EEF1D Mouse Monoclonal Antibody [clone: 3G4C4]
Antibodies
EEF1D, also named as EF1D and EF 1 delta, belongs to the EF-1-beta/EF-1-delta family. It is a subunit of the elongation factor-1 complex, which is responsible for the enzymatic delivery of aminoacyl tRNAs to the ribosome. EF-1-beta and EF-1-delta stimulate the exchange of GDP bound to EF-1-alpha to GTP. EEF1D is phosphorylated upon DNA damage, probably by ATM or ATR. This antibody is a mouse monoclonal antibody raised against residues near the C terminus of human EEF1D.

Western Blot:transfection MCF-7 Cells, 1:500-1:5000; IHC: Human pancreas cancer Tissue, 1:20-1:200

Type: Primary
Antigen: EEF1D
Clonality: Monoclonal
Clone: 3G4C4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR