Anti-DEPDC6 Rabbit Polyclonal Antibody

Supplier: Proteintech
20985-1-AP-150UL
10081-722EA 462.47 USD
10081-722
Anti-DEPDC6 Rabbit Polyclonal Antibody
Antibodies
DEPDC6, also named as DEPTOR, was identified as a novel mTOR-interacting protein whose over-expression in certain cell types represents a new mechanism for activating PI3K/Akt signaling and promoting cell survival. It negative regulator of the mTORC1 and mTORC2 signaling pathways. DEPDC6 inhibits the kinase activity of both complexes. The antibody is specific to DEPDC6.

Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human Liver cancer, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: DEPDC6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR