Anti-NFE2L1 Rabbit Polyclonal Antibody

Supplier: Proteintech
17062-1-AP-150UL
10081-668EA 462.47 USD
10081-668
Anti-NFE2L1 Rabbit Polyclonal Antibody
Antibodies
NFE2L1, also named as HBZ17, NRF1, TCF11 and LCR-F1, belongs to the bZIP family and CNC subfamily. It activates erythroid-specific, globin gene expression. NFE2L1 is involved in mediating ascorbic acid-induced osterix expression and osteoblast differentiation via binding to the antioxidant response element of the osterix promoter. NFE2L1, produced by osteoblasts, is involved in regulating osterix expression, osteoblast differentiation, and bone formation. . This antibody is a rabbit polyclonal antibody raised against residues near the C terminus of human ESRRG.

Western Blot: HEK-293 Cells, 1:200-1:2000

Type: Primary
Antigen: NFE2L1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR