Anti-NF45 Rabbit Polyclonal Antibody

Supplier: Proteintech
24572-1-AP-150UL
10081-664EA 462.47 USD
10081-664
Anti-NF45 Rabbit Polyclonal Antibody
Antibodies
Interleukin-2 enhancer binding factor 2 (ILF2), also named as nuclear factor 45 (NF45), plays important roles in regulating interleukin-2 expression in mammals. It forms a heterodimeric complex with ILF3, which may regulate transcription of the IL2 gene during T-cell activation. It can also promote the formation of stable DNA-dependent protein kinase holoenzyme complexes on DNA .

Western Blot:K-562 Cells, 1:500-1:5000; IHC: Human Colon Tissue, 1:20-1:200

Type: Primary
Antigen: NF45
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR