Anti-ILK Rabbit Polyclonal Antibody

Supplier: Proteintech
12955-1-AP-150UL
10081-570EA 462.47 USD
10081-570
Anti-ILK Rabbit Polyclonal Antibody
Antibodies
Integrin-linked kinase (ILK) is an ankyrin-repeat containing serine-threonine protein kinase capable of interacting with the cytoplasmic domains of integrin β1, β2, and β3 subunits and whose kinase activity is modulated by cell-extracellular matrix interactions. This protein belongs to the protein kinase superfamily. It can be autophosphorylated on serine residues.

Western Blot: Mouse Heart Tissue, 1:500-1:5000; IHC: Human pancreas cancer Tissue, 1:20-1:200

Type: Primary
Antigen: ILK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR