Anti-IDH2 Rabbit Polyclonal Antibody

Supplier: Proteintech
23254-1-AP-150UL
10081-548EA 462.47 USD
10081-548
Anti-IDH2 Rabbit Polyclonal Antibody
Antibodies
IDH2, also named as IDP and ICD-M, belongs to the isocitrate and isopropylmalate dehydrogenases family. It plays a role in intermediary metabolism and energy production. IDH2 is a mitochondrial NADP-dependent isocitrate dehydrogenase that catalyzes oxidative decarboxylation of isocitrate to alpha-ketoglutarate, producing NADPH. It may tightly associate or interact with the pyruvate dehydrogenase complex. IDH1 and IDH2 mutations could also contribute to tumorigenesis and cancer progression through increased mutagenesis.This antibody is specific to IDH2.

Western Blot: HEK-293 Cells, 1:500-1:5000; IHC: Human Heart, 1:20-1:200

Type: Primary
Antigen: IDH2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR