Anti-SPIN1 Rabbit Polyclonal Antibody

Supplier: Proteintech
19531-1-AP-150UL
10081-528EA 462.47 USD
10081-528
Anti-SPIN1 Rabbit Polyclonal Antibody
Antibodies
Spindlin 1 (SPIN1) is a stage specific embryonic protein. It was reported as an abundant maternal transcript present in the unfertilized egg and 2-cell, but not 8-cell, stage embryo. Spindlin 1 was phosphorylated in a cell cycle-dependent manner, undergoing posttranslational modification during the first meiotic cycle. This antibody is specific to spindlin 1 (SPIN1), not to other spindlins, such as SPIN2 and SPIN3.

Western Blot:SKOV-3 Cells, 1:500-1:5000; IHC: Human ovary tumor Tissue, 1:20-1:200; IF: Hela Cells, 1:10-1:100

Type: Primary
Antigen: SPIN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR