Anti-SMARCA4 Rabbit Polyclonal Antibody

Supplier: Proteintech
21634-1-AP-150UL
10081-514EA 462.47 USD
10081-514
Anti-SMARCA4 Rabbit Polyclonal Antibody
Antibodies
SMARCA4, also named as BAF190A, BRG1, SNF2B and SNF2L4, belongs to the SNF2/RAD54 helicase family. SMARCA4 is a transcriptional coactivator cooperating with nuclear hormone receptors to potentiate transcriptional activation. It is a component of the CREST-BRG1 complex, a multiprotein complex that regulates promoter activation by orchestrating a calcium-dependent release of a repressor complex and a recruitment of an activator complex. It is also involved in vitamin D-coupled transcription regulation via its association with the WINAC complex, a chromatin-remodeling complex recruited by vitamin D receptor (VDR), which is required for the ligand-bound VDR-mediated transrepression of the CYP27B1 gene.

Western Blot: HeLa Cells, 1:1000-1:10000; IHC: Human lung cancer Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100; IP: HeLa Cells, 1:500-1:5000

Type: Primary
Antigen: SMARCA4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR