Anti-SAR1A Rabbit Polyclonal Antibody

Supplier: Proteintech
22291-1-AP-150UL
10081-478EA 462.47 USD
10081-478
Anti-SAR1A Rabbit Polyclonal Antibody
Antibodies
SAR1A, also named as SAR1, SARA and SARA1, belongs to the small GTPase superfamily and SAR1 family. It is involved in transport from the endoplasmic reticulum to the Golgi apparatus. SAR1A is required to maintain SEC16A localization at discrete locations on the ER membrane perhaps by preventing its dissociation. SAR1A-GTP-dependent assembly of SEC16A on the ER membrane forms an organized scaffold defining endoplasmic reticulum exit sites (ERES). The antibody is specific to SAR1A.

Western Blot: HepG2 Cells, 1:200-1:2000; IHC: Human Skeletal Muscle Tissue, 1:20-1:200

Type: Primary
Antigen: SAR1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR