Anti-PRDX1 Rabbit Polyclonal Antibody

Supplier: Proteintech
20568-1-AP-150UL
10081-410EA 462.47 USD
10081-410
Anti-PRDX1 Rabbit Polyclonal Antibody
Antibodies
PRDX1, also named as PAGA, PAGB, TDPX2, PAG and NKEF-A, belongs to the ahpC/TSA family. It is involved in redox regulation of the cell. PRDX1 reduces peroxides with reducing equivalents provided through the thioredoxin system but not from glutaredoxin. PRDX1 plays an important role in eliminating peroxides generated during metabolism. PRDX1 might participate in the signaling cascades of growth factors and tumor necrosis factor-alpha by regulating the intracellular concentrations of H2O2. It reduces an intramolecular disulfide bond in GDPD5 that gates the ability to GDPD5 to drive postmitotic motor neuron differentiation. The antibody is specific to PRDX1.

Western Blot: HeLa Cells, 1:1000-1:10000; IF: HepG2, 1:10-1:100

Type: Primary
Antigen: PRDX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR